Visual colour kit, free chlorine
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1454201 1454202
HACH1454201EA 176 EUR
HACH1454201 HACH1454202
Visual colour kit, free chlorine
Milieutestkits
Simple, economical, rapid visual colour determination.
DPD visual test kit method for free chlorine, which can be used to accurately measure the free chlorine levels in the field or in the lab.
Leveringsinformatie: Kit includes one reagent bottle, stopper, colour comparator box, colour disc, tube, instruction sheet, and a carrying case.