Crucibles, quartz
Leverancier: CEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
303040 303060 303070
458-4225EA 0 EUR
458-4225 458-4226 458-4227
Crucibles, quartz
Kroezen
These quartz fibre crucibles reduce ashing times by as much as 97% and cool in seconds.
  • Withstand constant maximum temperature of 1000 °C and do not need desiccation