PEEK tubing
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AL35703 AL35716 AL35720 AL35722 AL35724 AL35725 AL35729 AL35741 AL35723 AL35709 AL35705 AL35708 AL35710 AL35711 AL35712 AL35714 AL35715 AL35718
VARIAL35703EA 0 EUR
VARIAL35703 VARIAL35716 VARIAL35720 VARIAL35722 VARIAL35724 VARIAL35725 VARIAL35729 VARIAL35741 VARIAL35723 VARIAL35709 VARIAL35705 VARIAL35708 VARIAL35710 VARIAL35711 VARIAL35712 VARIAL35714 VARIAL35715 VARIAL35718
PEEK tubing
Slangen Chromatography Tubing
Polymer tubing
  • For high-pressure HPLC plumbing
  • Maximum temperature: 100 °C