Accessories for data loggers, Dickson®
Leverancier: DICKSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A015 A016
DICKA015EA 146 EUR
DICKA015 620-1515
Accessories for data loggers, Dickson®
Dataloggers
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Order Now
Specificaties
Environmentally Preferable
Interface USB