Dropping bottles, LDPE, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WHEA211605EA 470 EUR
WHEA211605 WHEA211606 WHEA211603 WHEA211604
Dropping bottles, LDPE, WHEATON®
Flessen Druppelflessen
LDPE, translucent, with tip and cap.
  • Tip dispenses a stream of liquid or non-controlled drop
  • Flexible for easy handling
  • Good chemical and temperature resistance
Verpakking: Dropper bottles, tips and caps are packaged separately.