Total organic carbon (TOC) reagent sets, Test 'N Tube™
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2760345 2760445 2815945
705-0191EA 436 EUR
705-0191 705-0192 705-0196
Total organic carbon (TOC) reagent sets, Test 'N Tube™
Milieutestkits
Used for determination of Total Organic Carbon (TOC) either by low range, mid range or high range test 'N Tube' method respectively. The set includes digestion vials and reagents for 25 to 50 tests.
  • Affordable
  • Accurate
  • Easy to use
16 mm TNT products can be evaluated on DR900 only.