Nitrogen ammonia test kit, NI-8
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
224100 2428700
HACH224100EA 172 EUR
HACH224100 HACH2428700
Nitrogen ammonia test kit, NI-8
Milieutestkits
Simple, economical, rapid colour disc methods for testing ammonia in freshwater or salt water.
  • Most accurate visual test kit method
  • Simple, low cost colour disc method
  • Quick results with an easy-to-carry test kit