Mixing measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3212/04 3212/04WC 3212/06WC 3212/08 3212/08WC 3212/10 3212/10WC 3212/12WC 3212/14WC 3212/16WC 3212/18 3212/18WC
BIBB3212/04EA 71.2 EUR
BIBB3212/04 BIBB3212/04WC BIBB3212/06WC BIBB3212/08 BIBB3212/08WC BIBB3212/10 BIBB3212/10WC BIBB3212/12WC BIBB3212/14WC BIBB3212/16WC BIBB3212/18 BIBB3212/18WC
Mixing measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Cilinders Maatcilinders
Borosilicate glass, with polyethylene (PE) stopper, class A.
  • Cylinders have hexagonal foot
  • Graduations in white enamel for improved visibility with dark coloured solutions
Height values = maximum height
Certificaten: ISO 4788