Capillary columns, HP-1ms
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091S-602 19091S-612 19091S-633 19091S-713 19091S-733 19091S-833 19091S-911 19091S-913 19091S-916 19091S-931 19091S-933 19091S-933E 19091S-936 19091S-936E 19091S-602E 19091S-733E 19091S-913E 19091S-931E 19091S-633LTM 19091S-677 19091S-677E 19091S-677LTM 19091S-713LTM 19091S-733LTM 19091S-833LTM 19091S-933LTM 29091S-677LTM 29091S-833LTM 29091S-931LTM
558-0246EA 694 EUR
558-0246 HEWL19091S-612 HEWL19091S-633 HEWL19091S-713 HEWL19091S-733 HEWL19091S-833 HEWL19091S-911 HEWL19091S-913 HEWL19091S-916 HEWL19091S-931 HEWL19091S-933 HEWL19091S-933E HEWL19091S-936 HEWL19091S-936E HEWL19091S-602E HEWL19091S-733E HEWL19091S-913E HEWL19091S-931E HEWL19091S-633LTM HEWL19091S-677 HEWL19091S-677E HEWL19091S-677LTM HEWL19091S-713LTM HEWL19091S-733LTM HEWL19091S-833LTM HEWL19091S-933LTM HEWL29091S-677LTM HEWL29091S-833LTM HEWL29091S-931LTM
Capillary columns, HP-1ms
Kolommen voor chromatografie
These are excellent general purpose columns.
  • Low bleed characteristics
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded and cross-linked
100% Dimethylpolysiloxane, non-polar, solvent rinsable