Capillary columns, HP-5
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091J-002 19091J-005 19091J-012 19091J-015 19091J-101 19091J-102 19091J-105 19091J-112 19091J-112E 19091J-113 19091J-115 19091J-202 19091J-205 19091J-212 19091J-213 19091J-213E 19091J-215 19091J-216 19091J-231 19091J-233 19091J-236 19091J-313 19091J-333 19091J-411 19091J-413 19091J-413E 19091J-416 19091J-431 19091J-433 19091J-433E 19091J-436 19095J-023 19095J-023E 19095J-121 19095J-121E 19095J-123 19095J-123E 19095J-321 19095J-323 19095J-323E 19095J-621 19095J-623 19091J-436E 19091J-102E 19091J-012E 19091J-105E 19091J-113E 19091J-215E 19091J-216E 19091J-431E 19095J-623E 19091J-577 19091J-108 19091J-113LTM 19091J-413LTM 19091J-433LTM 19091J-577E 19091J-577LTM 19095J-121LTM
HEWL19091J-002EA 622 EUR
HEWL19091J-002 HEWL19091J-005 HEWL19091J-012 HEWL19091J-015 HEWL19091J-101 HEWL19091J-102 HEWL19091J-105 558-0239 HEWL19091J-112E 558-0390 HEWL19091J-115 HEWL19091J-202 558-0240 558-0391 HEWL19091J-213 HEWL19091J-213E HEWL19091J-215 HEWL19091J-216 HEWL19091J-231 HEWL19091J-233 HEWL19091J-236 HEWL19091J-313 HEWL19091J-333 HEWL19091J-411 558-0241 HEWL19091J-413E HEWL19091J-416 HEWL19091J-431 558-0242 HEWL19091J-433E 558-0243 558-0400 HEWL19095J-023E HEWL19095J-121 HEWL19095J-121E HEWL19095J-123 HEWL19095J-123E HEWL19095J-321 558-0401 HEWL19095J-323E HEWL19095J-621 HEWL19095J-623 HEWL19091J-436E HEWL19091J-102E HEWL19091J-012E HEWL19091J-105E HEWL19091J-113E HEWL19091J-215E HEWL19091J-216E HEWL19091J-431E HEWL19095J-623E HEWL19091J-577 HEWL19091J-108 HEWL19091J-113LTM HEWL19091J-413LTM HEWL19091J-433LTM HEWL19091J-577E HEWL19091J-577LTM HEWL19095J-121LTM
Capillary columns, HP-5
Kolommen voor chromatografie
These are excellent general purpose columns.
  • Non-polar
  • Wide range of applications
  • High temperature limit
  • Bonded and cross-linked
  • Solvent rinsable
  • Wide range of column dimensions available
(5%-Phenyl)-methylpolysiloxane