X-Type syringes for sensitive chromatography applications
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
204452 204475 204400 204379
549-0696EA 221 EUR
549-0696 549-0694 549-0695 HAMI204379
X-Type syringes for sensitive chromatography applications
Spuiten Injectiespuiten voor chromatografie
X-Type syringes are perfectly suited for sensitive LC samples such as proteins, peptides, phospholipids or smaller amines.
The syringe features a deactivated glass barrel and needle with an inorganic coating to prevent adsorption on the inner surfaces, resulting in nearly zero sample carryover. The coating creates a smooth inner barrel surface, leading to increased syringe lifetime by lowering friction during high speed applications while maintaining a perfect seal over the entire life of the syringe. The deactivated X-Type needle is directly attached to the barrel without using cement, further reducing the risk of sample carryover.
Fixed needle (FN) - This termination is found on CTC chromatography syringes. The unique design features a direct attachment of the needle to the barrel and eliminates contact between the sample solvent and the adhesive reducing carryover.
X-Type syringes are specially designed for the CTC LC PAL, HTC PAL, HTS PAL HTX PAL, PAL HTC-xt, PAL HTS-xt and PAL HTX-xt extended autosamplers sold under the following brands like AB Sciex, Agilent, Alpha M.O.S., Antek, Bruker, Dionex, GE, Gerstel, GL Sciences, Lauda, LEAP Technologies, MicroCal, MPS, Perichrom, Perkin Elmer, Shimadzu, Sotax, Thermo Scientific, Waters and Zoex.