Capillary columns, HP-INNOWAX
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091N-013 19091N-033 19091N-036 19091N-102 19091N-105 19091N-111 19091N-113 19091N-113E 19091N-116 19091N-131I 19091N-133 19091N-133E 19091N-136E 19091N-202 19091N-213 19091N-213E 19091N-216E 19091N-231I 19091N-233E 19095N-121E 19095N-123I 19095N-123E 19095N-126 19091N-030 19091N-105E 19091N-130 19091N-131E 19091N-205E 19091N-013LTM 19091N-030LTM 19091N-102LTM 19091N-133LTM 19091N-331 19091N-577I 19091N-577E 19091N-577LTM 19095N-123LTM 29091N-133LTM 29091N-577LTM
HEWL19091N-013EA 0 EUR
HEWL19091N-013 HEWL19091N-033 HEWL19091N-036 HEWL19091N-102 HEWL19091N-105 HEWL19091N-111 558-0392 HEWL19091N-113E HEWL19091N-116 HEWL19091N-131I 558-0393 HEWL19091N-133E HEWL19091N-136E HEWL19091N-202 HEWL19091N-213 HEWL19091N-213E HEWL19091N-216E HEWL19091N-231I HEWL19091N-233E HEWL19095N-121E HEWL19095N-123I HEWL19095N-123E HEWL19095N-126 HEWL19091N-030 HEWL19091N-105E HEWL19091N-130 HEWL19091N-131E HEWL19091N-205E HEWL19091N-013LTM HEWL19091N-030LTM HEWL19091N-102LTM HEWL19091N-133LTM HEWL19091N-331 HEWL19091N-577I HEWL19091N-577E HEWL19091N-577LTM HEWL19095N-123LTM HEWL29091N-133LTM HEWL29091N-577LTM
Capillary columns, HP-INNOWAX
Kolommen voor chromatografie
Recommended columns for alcohols, free organic acids, solvents, essential oils, flavors and fragrances.
  • High polarity
  • Highest upper temperature limits of the bonded PEG phases
  • Column-to-column repeatability
  • Bonded and cross-linked
Polyethylene glycol (PEG), solvent rinsable
Max. temperature: 250/270 °C