Capillary columns, DB-WAX
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121-7012 121-7022 121-7023 121-7043 122-7012 122-7013 122-7031 122-7032 122-7033 122-7061 122-7062 122-7063 123-7012 123-7013 123-7031 123-7032 123-7033 123-7033E 123-7062 123-7063 124-7032 125-7012 125-7017 125-7031 125-7032 125-7032E 125-7037 125-7062 126-7012 126-7013 127-7012 127-7012E 127-7013 127-7022 127-7023 128-7032 122-7032E 122-7033E 123-7032E 123-7063E 128-7022 128-7052 122-7062E 122-7063E 125-7012E 125-7062E 127-7023E 122-7012E 121-7012LTM 121-7022E 121-7022LTM 121-7023LTM 121-7042 121-7042E 122-7012LTM 122-7013LTM 122-7032LTM 122-7033LTM 123-7012LTM 123-7013LTM 123-7032LTM 123-7033LTM 125-7031LTM 125-7032LTM 127-7012LTM 127-7013LTM 127-7022LTM 127-7023LTM 128-7032LTM 222-7013LTM 222-7033LTM 122-7032UI
HEWL121-7012EA 397 EUR
HEWL121-7012 HEWL121-7022 HEWL121-7023 HEWL121-7043 HEWL122-7012 HEWL122-7013 HEWL122-7031 558-0367 HEWL122-7033 HEWL122-7061 HEWL122-7062 HEWL122-7063 HEWL123-7012 HEWL123-7013 HEWL123-7031 HEWL123-7032 558-0370 HEWL123-7033E HEWL123-7062 HEWL123-7063 HEWL124-7032 558-0373 HEWL125-7017 HEWL125-7031 558-0374 HEWL125-7032E HEWL125-7037 HEWL125-7062 HEWL126-7012 HEWL126-7013 HEWL127-7012 HEWL127-7012E HEWL127-7013 HEWL127-7022 HEWL127-7023 HEWL128-7032 HEWL122-7032E HEWL122-7033E HEWL123-7032E HEWL123-7063E HEWL128-7022 HEWL128-7052 HEWL122-7062E HEWL122-7063E HEWL125-7012E HEWL125-7062E HEWL127-7023E HEWL122-7012E HEWL121-7012LTM HEWL121-7022E HEWL121-7022LTM HEWL121-7023LTM HEWL121-7042 HEWL121-7042E HEWL122-7012LTM HEWL122-7013LTM HEWL122-7032LTM HEWL122-7033LTM HEWL123-7012LTM HEWL123-7013LTM HEWL123-7032LTM HEWL123-7033LTM HEWL125-7031LTM HEWL125-7032LTM HEWL127-7012LTM HEWL127-7013LTM HEWL127-7022LTM HEWL127-7023LTM HEWL128-7032LTM HEWL222-7013LTM HEWL222-7033LTM HEWL122-7032UI
Capillary columns, DB-WAX
Kolommen voor chromatografie
These columns improve resolution of low boiling point analytes.
  • High polarity
  • Lower temperature limit of 20 °C
  • Column-to-column reproducibility
  • Bonded and cross-linked
  • Solvent rinsable
Polyethylene glycol (PEG)
Order Now
Specificaties
Environmentally Preferable