Microcentrifuge tubes
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72.706.600 72.704.700 72.706.700
SARS72.706.600EA 0 EUR
SARS72.706.600 SARS72.704.700 SARS72.706.700
Microcentrifuge tubes
Buisjes Microcentrifugebuisjes
PP, transparent.
  • Wide hinge for easy and precise closure
  • Large writing space on frosted, flat lid
  • DNA free, DNase/RNase free
  • PCR inhibitor free
The low protein and DNA binding graduated micro tubes with SafeSeal locking cap minimises the sample loss.