Detectiereagens voor western blotting, Amersham ECL™ Start
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RPN3244 RPN3243
RPN3244EA 244 EUR
RPN3244 RPN3243
Detectiereagens voor western blotting, Amersham ECL™ Start
Reagentia voor elektroforese Western Blotting Reagents
Chemiluminescence detection reagent for detection of high to medium abundant proteins.
  • For outine and confirmatory Western blotting, high expressed proteins and tagged proteins
  • Detection with X-ray film or CCD Imager
  • Stable until the expiration date printed on box when stored under recommended conditions
Recommended detection with Amersham Hyperfilm™ (X-ray film) or ImageQuant™ LAS 500. Optimal membrane Amersham Protran.
Order Now
Specificaties
Environmentally Preferable