Adapter for exhaust filter for Justrite and CPC canisters
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCTV107610EA 39.9 EUR
SCTV107610 SCTV160527 SCTV160529 SCTV318957 SCTV107617 SCTV108181 SCTV160524
Adapter for exhaust filter for Justrite and CPC canisters
Fittingen en connectoren
Special adapter for connecting a SCAT exhaust air filter with the canisters of the manufacturers Justrite and CPC.
  • Made of PEHD-EC material
  • Fits GL 14 (f) thread on CPC (m) coupling
Compatible for all SCAT exhaust filters sizes small, medium, and large.