Transport trolleys
Leverancier: HUPFER METALLWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7500468 7500661
139-0771EA 583 EUR
139-0771 139-0772
Transport trolleys
Transportwagens
Transport trolley with high quality stainless steel construction, complete with stable tubes of 25×25 mm, and two smooth welded shelves.
  • Zinc-chrome plate steel housing, anti-static with bumpers
  • Chassis complete with four swivel castors, front two lockable