Heating mantles, multi-position, WHRE6 series
Leverancier: WINKLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WHRE6E000500RC WHRE6E000100CH WHRE6E000100RC WHRE6E000100SS WHRE6E000250CH WHRE6E000250RC WHRE6E000500CH WHRE6E000500SS WHRE6E001000CH WHRE6E001000RC WHRE6E001000SS WHRE6E000250SS WHRE6E000100UK WHRE6E000250UK WHRE6E000500UK WHRE6E001000UK
WINKWHRE6E000500RCEA 2555 EUR
WINKWHRE6E000500RC WINKWHRE6E000100CH WINKWHRE6E000100RC WINKWHRE6E000100SS WINKWHRE6E000250CH WINKWHRE6E000250RC WINKWHRE6E000500CH WINKWHRE6E000500SS WINKWHRE6E001000CH WINKWHRE6E001000RC WINKWHRE6E001000SS WINKWHRE6E000250SS WINKWHRE6E000100UK WINKWHRE6E000250UK WINKWHRE6E000500UK WINKWHRE6E001000UK
Heating mantles, multi-position, WHRE6 series
Verwarmers Verwarmingsmantels
For round bottomed flasks. Max. 450 °C. Multiposition heating mantles with powder coated, chemically resistant metal housing and stainless steel front and cover plate. Mantles have crocheted outer jacket made from glass silk.
  • Integrated power selector per heating position, infinitely variable power 0 to 100%
  • Six heating zones
  • Detachable plug-in connection (cable length 0,5 m)
Leveringsinformatie: Supplied without clamping rod, rods are available as an optional accessory.