Lab Consumables
Leverancier: Copan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
310C25T 311C25T 312C25T 313C25T 316C05T 317C05T 318C05T 319C05T K34 K108 K44 K45 K46 K47 214C 175CS01 176CS01 177CS01 178CS01 179CS01
612-2668EA 190 EUR
612-2668 612-2669 612-2670 612-2671 612-2674 612-2675 612-2676 612-2677 612-2681 612-2682 612-2683 612-2684 612-2685 612-2686 COPA214C COPA175CS01 COPA176CS01 COPA177CS01 COPA178CS01 COPA179CS01
Lab Consumables
Uitstrijkspatels Entogen en -naalden
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification