Hand cleansing creams, Kresto® paint liquid
Leverancier: SC Johnson
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Kresto®
EVON22306EA 231 EUR
EVON22306
Hand cleansing creams, Kresto® paint liquid
Huidverzorging Zepen en huidreinigingsmiddelen
The special hand cleanser with very powerful cleansing action contains mild solvents. It removes very stubborn and extremely heavy-duty contamination like lacquers, resins, adhesives, oil-based paints.
  • Scrubbing agent-free
  • As the formulation is concentrated, only a small amount is required to remove contaminants
  • Solvent-free, colourant-free, preservative-free, perfumed
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904