Storage Boxes
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5973 5971
SEMA5973EA 70.8 EUR
SEMA5973 SEMA5971
Storage Boxes
Dozen Opbergdozen
PP, translucent.
  • Very stable
  • Good chemical resistance
  • Good temperature resistance
Multipurpose boxes for a wide range of uses.