General purpose trays
Leverancier: Gilac PROFESSIONNEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G179321 G179521 G179621 G179721
CORAG179321EA 35.8 EUR
CORAG179321 CORAG179521 CORAG179621 CORAG179721
General purpose trays
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Trays, made of HDPE, ideal for lifting the dough and used for storage purpose.
  • High quality and durability
  • Good chemical resistance
  • Resist temperature from –40 to +90 °C