Tubings, insulated
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RKJ016 RKJ041 RKJ059 RKJ111 RKJ112
LAUDRKJ016EA 44.1 EUR
LAUDRKJ016 LAUDRKJ041 LAUDRKJ059 LAUDRKJ111 LAUDRKJ112
Tubings, insulated
Slangen Transfer Tubing
Polymer tubing, EPDM or silicone
  • EPDM tube is not for Ultra 350 and mineral oils
  • Silicone tubes should not used in conjunction with silicone oils
Temperature resistance: +10 to +90 °C (EPDM), +10 to +100 °C (silicone).