Precisiethermometer
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M235000-FL/VC M235000-FL M235002-FL M235004-GA
AMARM235000-FL/VCEA 319 EUR
AMARM235000-FL/VC AMARM235000-FL AMARM235002-FL AMARM235004-GA
Precisiethermometer
Thermometers Analoge thermometers
Dropping point thermometers according to Ubbelohde.
  • Interne schaalverdeling in melkglas
  • Ronde Richter-afwerking aan bovenkant
  • Kleurloos, prismatisch capillair
These enclosed milk glass scale thermometers are suitable for government verification or for government calibration.
Certificaten: Conform DIN 12785
Bestelinformatie: Available with or without verification certificate.
Verpakking: Packed in a transparent square plastic case.