Tweezers, NANO
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00-NANO 0-NANO 1-NANO 2A-NANO 3C-NANO SS-NANO 3-NANO 5A-NANO 5-NANO 7-NANO
232-0148EA 35.8 EUR
232-0148 232-0157 232-0160 232-0163 232-0166 232-0181 232-0169 232-0172 232-0175 232-0178
Tweezers, NANO
Pincetten
Ultra-modern nanotechnology closes even the tiniest of pores on the surface of the Nano tweezers. The instruments are made from extremely high grade surgical stainless steel. The Lotus effect, engineered by the nanotechnology, causes dirt, oil, water and other liquids to simply pearl off the steel. This keeps the tweezers perfectly clean, a beneficial feature particularly in a sensitive laboratory setting.
  • Very clean tweezer surface
  • Ideal for use in cleanrooms and other sensitive laboratory areas where contaminant-free tools are required