Scoops, Open PharmaScoop®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A336-1000 A336-250 A336-500 A336-750 A336-2000 A336-1500
SAMPA336-1000EA 171 EUR
SAMPA336-1000 SAMPA336-250 SAMPA336-500 SAMPA336-750 SAMPA336-2000 SAMPA336-1500
Scoops, Open PharmaScoop®
Scheppen
These general purpose scoops made from stainless steel 316 feature a heavy duty, crevice free construction which makes them ideal for use in the food and pharmaceutical industries.
  • Open top design, flat bottom with rounded edges
  • 316 stainless steel is a high-quality grade and is therefore very corrosion resistant
  • Satin finish