Taps for aspirator bottles
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02801904 02802906
271-9001EA 53.1 EUR
271-9001 271-9002
Taps for aspirator bottles
Kranen en gootstenen Kranen
These drain taps are made of borosilicate glass and are used for aspirator bottles.