Gas washing bottles heads
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05530301 05530300
LENZ05530301EA 49.5 EUR
LENZ05530301 LENZ05530300
Gas washing bottles heads
Sluitingen Flesdoppen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes