Tissue grinders, DUALL®, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP885450-0020 SP885450-0021 SP885450-0022 SP885450-0023 SP885450-0024 SP885450-0025
SCERSP885450-0020EA 288 EUR
SCERSP885450-0020 SCERSP885450-0021 SCERSP885450-0022 SCERSP885450-0023 SCERSP885450-0024 SCERSP885450-0025
Tissue grinders, DUALL®, KIMBLE®
Homogenisatoren Potterbuizen
These borosilicate glass tissue grinders combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Construction is strong enough to allow the pestle to be motor driven and also the grinding efficiency is greatly improved while using this tube.
  • Replacement components are available and completely interchangeable
  • Used for connective tissues such as muscle, heart and lung
Clearance between pestles and tubes is 0,004 to 0,006"