Water sampling bottles, PP, wide neck
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
411-0125 411G10R27
EPSC411-0125EA 495 EUR
EPSC411-0125 EPSC411G10R27
Water sampling bottles, PP, wide neck
Flessen Watermonsternameflessen
PP, sterile, with leak-proof PP closure
  • Wide mouth for easy aseptic filling
  • Leakproof and break resistant
  • Each container individually barcoded with lot number and unique container number
  • Each closure individually sealed with tamper-evident shrink wrap
These are recommended for collecting water samples for microbiological analyses.
Leveringsinformatie: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Verpakking: Carton sealed with security tape providing tamper evidence. The chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping.