Dropping bottles, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F116600000 F116610000 F116620000
BELAF116600000EA 67.8 EUR
BELAF116600000 BELAF116610000 BELAF116620000
Dropping bottles, SP Bel-Art
Flessen Druppelflessen
LDPE, translucent.
  • Good chemical resistance, heat resistant up to +80 °C
  • Good temperature resistance
  • Very flexible for easy handling
Ideal for staining slides, indicator solutions or dyes. It can be used as squirt bottles with the supplied tube and as dropper bottles when the tube is removed.