Racks for centrifuge tubes, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F187950001 F187950000
BELAF187950001EA 66.5 EUR
BELAF187950001 BELAF187950000
Racks for centrifuge tubes, SP Bel-Art
Rekken Tube Racks
PS, coloured
  • Non grip style allows the tube to be lifted freely from holder
  • Grip style holds the tube which can be removed by holding the base