Containers, wide-mouth, straight, with cap
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
311-1000 311-0500
216-0376EA 131 EUR
216-0376 216-0392
Containers, wide-mouth, straight, with cap
Monsternamepotjes Containers voor algemeen gebruik
These straight sided wide-mouth HDPE jar allows for easy filling and sample retreival. Recommended for collecting soil, sediment and sludge samples to be analyzed for inorganic trace analytes.
  • With white LDPE-lined PP cap
  • Caps assembled onto containers
  • Individually barcoded with lot number and unique container number
Certificaten: Certified to meet or exceed EPA perfomance-based standards for metals, cyanide and fluoride.
Leveringsinformatie: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement