Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Leverancier: Bel-Art Products, a part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Odo-Clave®
BELAH131990000EA 455 EUR
BELAH131990000
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Deodorants en luchtverfrissers
This heat-released deodorant pads neutralise autoclave odors with a pleasant scent.
  • Simply open foil pouch and place pad in bag prior to autoclaving
  • Pads have a three year life span; will last longer if stored in a sealed container
  • Triple strength clove releases three times the deodorant of the standard pads while costing less than two standard pads
W×L: 57×57 mm
Verpakking: Packed 10 pads per bag, 10 bags per box.