Wash bottles, narrow neck, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F116210008 F116210016
BELAF116210008EA 269 EUR
BELAF116210008 BELAF116210016
Wash bottles, narrow neck, SP Bel-Art
Flessen Spuitflessen
LDPE, with PP cap und delivery tube
  • Simple to squeeze
  • Extra fine needle spray
  • Good chemical resistance, good temperature resistance
Ultra thin spray for precise aiming.