TLC plates, kieselguhr and mixed layers
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.05567.0001 1.05568.0001 1.05738.0001
1.05567.0001EA 214 EUR
1.05567.0001 1.05568.0001 1.05738.0001
TLC plates, kieselguhr and mixed layers
TLC-platen
Kieselguhr is a natural diatomaceous earth that can be used for the separation of polar or moderately polar compounds. For the mixed layer plates a combination of classical silica gel 60 and kieselguhr is used, providing favorable properties for certain applications such as separations of inorganic ions, herbicides or steroids.