Measuring cylinder, tall form, class B, PP
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7503-0100 7503-0050 7503-0250 7503-0500 7503-1000 7503-2000
BURK7503-0100EA 9.7 EUR
BURK7503-0100 BURK7503-0050 BURK7503-0250 BURK7503-0500 BURK7503-1000 BURK7503-2000
Measuring cylinder, tall form, class B, PP
Cilinders Maatcilinders
PP, highly transparent, tall form, class B.
  • Embossed blue graduation with rings at the main points
  • Shatterproof at room temperature
  • With spout and hexagonal base
  • Adjusted to 'In'
Certificaten: DIN 12681 / ISO 6706
Order Now
Specificaties
Environmentally Preferable