HPLC columns, SUPELCOSIL™, LC-NH₂, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
58338 59558 59568 59568C30 58338C30 58338C40 58988 58989 58989C30
58338.EA 831 EUR
58338. 59558. 59568. 59568C30 58338C30 58338C40 58988. 58989. 58989C30
HPLC columns, SUPELCOSIL™, LC-NH₂, Supelco®
Kolommen voor chromatografie
Silica based SUPELCOSIL™ LC-NH₂ columns separate monosaccharides, disaccharides, and some trisaccharides, with elution generally in order of increasing molecular weight. Retention decreases as the proportion of water:acetonitrile in the mobile phase is increased.
The amino column is most often employed for the separation of mono- and disaccharides.