Disposable filters, Optimizer®
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CWAZ0S1S3 CWAG0S1S3 CWMX0S0X1 CWMX0S1S3 CWNT3L0F1 CWAV0S1S3 CWAT0S1S3M CWAY0S1HC CWAZ0S1HC CWSF0S2S3M CWST0S2S3M
ENTECWAZ0S1S3EA 0 EUR
ENTECWAZ0S1S3 ENTECWAG0S1S3 ENTECWMX0S0X1 ENTECWMX0S1S3 ENTECWNT3L0F1 ENTECWAV0S1S3 ENTECWAT0S1S3M ENTECWAY0S1HC ENTECWAZ0S1HC ENTECWSF0S2S3M ENTECWST0S2S3M
Disposable filters, Optimizer®
Filters Filterpatronen
Photochemical filter with superior contaminant retention and low hold-up volume performance in sub-10 to 180 nm technology nodes.