HPLC columns, Hibar®, with ChiraDex® sorbents
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.00423.7004 1.00424.7004 1.50009.7004 1.50012.7004 1.50013.7004 1.50182.7004 1.50183.7004
1.00423.7004EA 0 EUR
1.00423.7004 1.00424.7004 1.50009.7004 1.50012.7004 1.50013.7004 1.50182.7004 1.50183.7004
HPLC columns, Hibar®, with ChiraDex® sorbents
Kolommen voor chromatografie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with Chiradex® sorbents. Ideal for separation of enantiomers of different class of substances.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.