Cell culture roller bottles, DURAN®
Leverancier: Carl Roth
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROTHT967.1EA 0 EUR
ROTHT967.1
Cell culture roller bottles, DURAN®
Flessen Rolflessen
DURAN®, borosilicate glass with DIN thread (GL 45)
  • Very good temperature resistance
  • Very good chemical resistance
Supplied with blue PP screw cap and pouring ring.