Peqlab peqGOLD, 8 PCR Cap Strips
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Peqlab
732-3224EA 54.7 EUR
732-3226 732-3205 732-3224
Peqlab peqGOLD, 8 PCR Cap Strips
Buisjes PCR-buisjes
PP, colourless. Caps ready for use with eight tube strips or 96 well PCR plates available in flat, domed or with ultra-transparent and minimal autofluorescent properties ideal for Real-Time PCR.
  • Certified free from RNase, DNase and human DNA
  • Manufactured from virgin polypropylene under strict quality control guidelines
Domed cap strips
Suitable for all standard PCR applications.
Flat cap strips
For easy tube marking and maximum contact with the thermal cycler’s heated lid.
QPCR cap strips
Highest level of optical transparency and minimum level of autofluorescence making these tubes ideal for Real-Time PCR.