Antimicrobial incise drapes, Ioban™ 2
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M6635 M6650 6640EU
MMMEM6635EA 165 EUR
MMMEM6635 MMMEM6650 MMME6640EU
Antimicrobial incise drapes, Ioban™ 2
Eerste-hulpbenodigdheden Operatielakens
These antimicrobial incise drapes provide a sterile surface to the wound edge at the start of surgery and continuous antimicrobial activity throughout the procedure. Ioban 2 film helps reduce the risk of surgical site contamination due to skin flora by providing a physical barrier to skin bacteria. The film adheres securely to wound edges which is critical in maintaining the barrier to skin flora.