Disposable earplugs, uvex xact-fit
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2124.003 2124.002 2124.001
111-1691EA 72.4 EUR
111-1691 111-1690 111-1689
Disposable earplugs, uvex xact-fit
Oordoppen Oordoppen voor eenmalig gebruik
These innovative earplugs adapt perfectly to the shape of the ear and feature a revolutionary design which imitates the anatomy of each ear. This noticeably reduces the pressure in the ear canal, even when worn for long periods. Earplugs do not need to be rolled, which allows for more hygienic handling.
  • Ergonomically-shaped grip for the left and right ears increases wearer comfort
  • Quick and easy insertion
  • Adjustable cord
SNR: 26 dB
EN 352-2:2002
Certificaten: EN 352-2:2002
Bestelinformatie: Also available as a refill box for the wall-mountable 'one2click' dispenser.