Coating thickness gauges, PosiTest
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LD5004 LD5003
SIMELD5004EA 524 EUR
SIMELD5004 SIMELD5003
Coating thickness gauges, PosiTest
Coatinglaag testers
These thickness gauges are designed for the measurement of coatings of painting, enamel, galvanizing, metalising and plating on steel.
  • Visual and audible indicators when thickness ready
  • V grooves in probe housing and gauge base
  • Correct positioning on cylindrical objects