Wafer shipping boxes, Ultrapak® series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H9200-0022 H9200-0023
ENTEH9200-0022EA 0 EUR
ENTEH9200-0022 ENTEH9200-0023
Wafer shipping boxes, Ultrapak® series
Wafertransportsystemen Wafer verzendcontainers
Wafershield™ high purity PP
  • Horizontal and vertical robotic pickup flanges on the cassettes
  • Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
Shipper consists of individual horizontal cantilever springs, which limits wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage.
Order Now
Specificaties
Environmentally Preferable