Fractionating columns, plain, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FC7/43 FC7/23
BIBBFC7/43EA 62.2 EUR
BIBBFC7/43 201-3033
Fractionating columns, plain, Quickfit®
Onderdelen voor destillatie Destillatiekolommen
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ground glass joints for dependable performance. Designed for use with various types of column packing e.g. Raschig rings or Fenske helices.