Multiple adapters, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MA2/2 MA2/3
201-3000EA 72.4 EUR
201-3000 201-3001
Multiple adapters, Quickfit®
Onderdelen voor destillatie Destillatieadapters met meerdere halzen
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two necks. One vertical and one angled at 45°.