Spin columns, PAGE-Optimizer™
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-947 786-948
786-947EA 91.3 EUR
786-947 786-948
Spin columns, PAGE-Optimizer™
Filters Centrifugefilters
PAGE-Optimizer™ is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents. PAGE-Optimizer™ helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer™ has also been shown to reveal protein bands previously masked by the above interfering agents.
  • Eliminate smeared lanes
  • Reveal key missing protein bands
  • Resolve distorted bands for improved visualization
  • Prevent overlapping lanes resulting in contamination and distortion ofadjacent lanes
  • Reduce swollen lanes to prevent protein contamination