Gown racks for frocks and lab coats
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GR32CMP-S GR40CMP-S GR08CMP-S-C GR16CMP-S-C GR24CMP-S-C GR40CMP-1750MM GR08CMP-1750MM GR32CMP-1750MM GR16CMP-S GR08CMP-S GR24CMP-S
PALBGR32CMP-SEA 2550 EUR
PALBGR32CMP-S PALBGR40CMP-S 132-0985 132-0986 132-0987 132-1086 PALBGR08CMP-1750MM PALBGR32CMP-1750MM 132-1085 PALBGR08CMP-S PALBGR24CMP-S
Gown racks for frocks and lab coats
Meubilair Omkleedrekken
These shorter gown racks are designed for hanging lab coats and frocks.
  • Rack design holds each hanger at exactly the same angle and spacing ensuring a uniform appearance, creating an image of orderliness and allowing sufficient laminar flow
  • Optional mirrors (purchased separately) can be connected to side panels, enhancing both aesthetics and functionality
  • Each gowning rack is supplied with hangers, labelling numbers and leveller legs with integral covers
  • Hangers - the thickness of the material ensures non deforming
  • Supplied with leveller legs or cleanrooms